6-Bromo-o-toluonitrile ≥97%

Supplier: AOBChem USA

Warning

27401-5G 27401-25G
77361-200EA 125.66 USD
77361-200 77372-962
6-Bromo-o-toluonitrile ≥97%
6-Bromo-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
Storage Temperature: Ambient
MDL Number: MFCD09834775
CAS Number: 77532-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR