5-Bromo-2-methoxyphenyl acetate ≥95%

Supplier: AOBChem USA

Warning

26050-5G 26050-25G
77361-080EA 142.14 USD
77361-080 77372-842
5-Bromo-2-methoxyphenyl acetate ≥95%
5-Bromo-2-methoxyphenyl acetate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27997554
CAS Number: 66037-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR