2-Methoxy-6-(methylsulfanyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25805-5G 25805-25G
77361-042EA 131.84 USD
77361-042 77372-814
2-Methoxy-6-(methylsulfanyl)benzaldehyde ≥97%
2-Methoxy-6-(methylsulfanyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
Storage Temperature: Ambient
MDL Number: MFCD18905252
CAS Number: 67853-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR