Potassium-2-(aminocarbonyl)phenyltrifluoroborate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Potassium (2-carbamoylphenyl)trifluoroborate

25555-5G 25555-25G
77361-008EA 176.13 USD
77361-008 77372-772
Potassium-2-(aminocarbonyl)phenyltrifluoroborate ≥97%
Potassium 2-(aminocarbonyl)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₆BF₃KNO
Storage Temperature: Ambient
MDL Number: MFCD04115765
CAS Number: 850623-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR