1-Chloro-3-fluoro-2-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

25163-5G 25163-25G
77360-964EA 121.54 USD
77360-964 77372-734
1-Chloro-3-fluoro-2-(methoxymethoxy)benzene ≥95%
1-Chloro-3-fluoro-2-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619419
CAS Number: 1865062-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR