2-Bromo-3,5-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

25134-5G 25134-25G
77360-962EA 113.3 USD
77360-962 77372-732
2-Bromo-3,5-dimethylphenol ≥97%
2-Bromo-3,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22058629
CAS Number: 125237-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR