5-Bromo-3-chloro-2-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

24862-5G 24862-25G
77360-924EA 173.04 USD
77360-924 77372-712
5-Bromo-3-chloro-2-(methylthio)pyridine ≥95%
5-Bromo-3-chloro-2-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNS
MW: 238.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29916541
CAS Number: 261625-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR