5-Bromo-6-chloropyridin-3-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Bromo-2-chloro-5-hydroxypyridine
24464-5G
24464-25G
77360-880EA
84.46
USD
77360-880
77372-682
5-Bromo-6-chloropyridin-3-ol ≥95%
5-Bromo-6-chloropyridin-3-ol
Precaución: for Research Use Only
Formula:
C₅H₃BrClNO MW: 208.45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835248 CAS Number: 130115-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...