1-Bromo-4-(2-methoxyethoxy)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
23903-5G
23903-25G
77360-820EA
115.36
USD
77360-820
77372-648
1-Bromo-4-(2-methoxyethoxy)benzene ≥97%
1-Bromo-4-(2-methoxyethoxy)benzene
Precaución: for Research Use Only
Formula:
C₉H₁₁BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09931307 CAS Number: 39255-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...