4-Bromo-2-fluorobenzaldoxime ≥97%

Supplier: AOBChem USA

Warning

23205-5G 23205-25G
77360-750EA 139.05 USD
77360-750 77372-604
4-Bromo-2-fluorobenzaldoxime ≥97%
4-Bromo-2-fluorobenzaldoxime

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD00143262
CAS Number: 202865-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR