2,6-Dichloro-4-nitrotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3-Dichloro-2-methyl-5-nitrobenzene

22715-5G 22715-25G
77360-730EA 77.25 USD
77360-730 77372-586
2,6-Dichloro-4-nitrotoluene ≥97%
2,6-Dichloro-4-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174267
CAS Number: 7149-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR