2-Bromo-4-methyl-1-(methylthio)benzene ≥97%

Supplier: AOBChem USA

Warning

21938-5G 21938-25G
77360-672EA 173.04 USD
77360-672 77372-532
2-Bromo-4-methyl-1-(methylthio)benzene ≥97%
2-Bromo-4-methyl-1-(methylthio)benzene

Precaución: for Research Use Only

Formula: C₈H₉BrS
MW: 217.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527261
CAS Number: 89981-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR