2,4-Dichloro-1-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

21346-5G 21346-25G
77360-628EA 90.64 USD
77360-628 77372-500
2,4-Dichloro-1-(methoxymethoxy)benzene ≥95%
2,4-Dichloro-1-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O₂
MW: 207.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28101427
CAS Number: 827-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR