5-Fluoro-2-formylpyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluoropyridine-2-carboxaldehyde, 5-Fluoropicolinaldehyde

21282-5G 21282-25G
77360-612EA 78.28 USD
77360-612 77372-490
5-Fluoro-2-formylpyridine ≥95%
5-Fluoro-2-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄FNO
MW: 125.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781242
CAS Number: 31181-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR