6-Bromo-5-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

21025-5G 21025-25G
77360-592EA 173.04 USD
77360-592 77372-480
6-Bromo-5-methyl-1H-indazole ≥97%
6-Bromo-5-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026997
CAS Number: 1000343-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR