2-Bromo-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

19929-5G 19929-25G
77360-528EA 78.28 USD
77360-528 77372-436
2-Bromo-3-iodopyridine ≥97%
2-Bromo-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃BrIN
Storage Temperature: Ambient
MDL Number: MFCD09056782
CAS Number: 265981-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR