3,5-dimethoxyphenyl diethylcarbamate ≥97%

Supplier: AOBChem USA

Warning

19406-5G 19406-25G
77360-498EA 131.84 USD
77360-498 77372-412
3,5-dimethoxyphenyl diethylcarbamate ≥97%
3,5-dimethoxyphenyl diethylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₄
MW: 253.294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755740
CAS Number: 1025324-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR