5-Bromothiophene-2-carboxaldoxime ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromothiophene-2-carbaldehyde oxime

19086-5G 19086-25G
77360-480EA 103 USD
77360-480 77372-406
5-Bromothiophene-2-carboxaldoxime ≥97%
5-Bromothiophene-2-carboxaldoxime

Precaución: for Research Use Only

Formula: C₅H₄BrNOS
MW: 206.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00126375
CAS Number: 2160-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR