Benzyl 2,4-difluorophenyl ether ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-(Benzyloxy)-2,4-difluorobenzene
18972-5G
18972-25G
77360-474EA
100.94
USD
77360-474
77372-400
Benzyl 2,4-difluorophenyl ether ≥97%
Benzyl 2,4-difluorophenyl ether
Precaución: for Research Use Only
Formula:
C₁₃H₁₀F₂O MW: 220.215 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12198257 CAS Number: 152434-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...