N-(4-Bromo-2,6-dimethylphenyl)-2,2,2-trifluoroacetamide ≥97%

Supplier: AOBChem USA

Warning

18914-5G 18914-25G
77360-472EA 90.64 USD
77360-472 77372-398
N-(4-Bromo-2,6-dimethylphenyl)-2,2,2-trifluoroacetamide ≥97%
N-(4-Bromo-2,6-dimethylphenyl)-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD07175852
CAS Number: 872415-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR