2,4-Dibromo-5-fluorotoluene ≥97%

Supplier: AOBChem USA

Warning

18617-5G 18617-25G
77360-446EA 100.94 USD
77360-446 77372-376
2,4-Dibromo-5-fluorotoluene ≥97%
2,4-Dibromo-5-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
Storage Temperature: Ambient
MDL Number: MFCD07368760
CAS Number: 93765-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR