4-Methoxy-2,5-dimethylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

18486-5G 18486-25G
77360-440EA 142.14 USD
77360-440 77372-368
4-Methoxy-2,5-dimethylphenylboronic acid ≥95%
4-Methoxy-2,5-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD06801676
CAS Number: 246023-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR