N,N-Diisopropyl-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

18031-5G 18031-25G
77360-414EA 167.89 USD
77360-414 77372-346
N,N-Diisopropyl-4-methylbenzamide ≥97%
N,N-Diisopropyl-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO
Storage Temperature: Ambient
MDL Number: MFCD00448125
CAS Number: 6937-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR