4-Bromo-N,N-3,5-tetramethylaniline ≥97%

Supplier: AOBChem USA

Warning

18007-5G 18007-25G
77360-412EA 131.84 USD
77360-412 77372-344
4-Bromo-N,N-3,5-tetramethylaniline ≥97%
4-Bromo-N,N-3,5-tetramethylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN
Storage Temperature: Ambient
MDL Number: MFCD11036245
CAS Number: 14275-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR