4-Bromo-1-methyl-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

17854-5G 17854-25G
77360-406EA 73.13 USD
77360-406 77372-338
4-Bromo-1-methyl-1H-indazole ≥95%
4-Bromo-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09870042
CAS Number: 365427-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR