5-Bromo-N-cyclohexylpicolinamide ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-N-cyclohexylpyridine-2-carboxamide

17383-5G 17383-25G
77360-384EA 162.74 USD
77360-384 77372-318
5-Bromo-N-cyclohexylpicolinamide ≥95%
5-Bromo-N-cyclohexylpicolinamide

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09801063
CAS Number: 951885-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR