3-Bromo-4-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

164686-5G 164686-25G
77360-320EA 127.72 USD
77360-320 77372-276
3-Bromo-4-ethoxybenzoic acid ≥97%
3-Bromo-4-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD01143606
CAS Number: 24507-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR