Ethyl(4-fluorophenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

161542-5G 161542-25G
77360-292EA 156.56 USD
77360-292 77372-252
Ethyl(4-fluorophenyl)sulfane ≥95%
Ethyl(4-fluorophenyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₉FS
Storage Temperature: Ambient
CAS Number: 2924-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR