2-Bromo-6-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

15557-5G 15557-25G
77360-244EA 172.01 USD
77360-244 77372-206
2-Bromo-6-(trifluoromethyl)benzoic acid ≥97%
2-Bromo-6-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06253757
CAS Number: 177420-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR