N,N-Dibenzyl-5-bromopyrimidin-2-amine ≥97%

Supplier: AOBChem USA

Warning

15029-5G 15029-25G
77360-198EA 131.84 USD
77360-198 77372-160
N,N-Dibenzyl-5-bromopyrimidin-2-amine ≥97%
N,N-Dibenzyl-5-bromopyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₁₈H₁₆BrN₃
Storage Temperature: Ambient
MDL Number: MFCD03648497
CAS Number: 886366-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR