(6-(Methoxycarbonyl)pyridin-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

14967-5G 14967-25G
77360-194EA 165.83 USD
77360-194 77372-156
(6-(Methoxycarbonyl)pyridin-3-yl)boronic acid ≥97%
(6-(Methoxycarbonyl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₄
MW: 180.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800559
CAS Number: 1072945-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR