1-Bromo-1-methoxy-4-phenoxybenzene ≥97%

Supplier: AOBChem USA

Warning

14916-5G 14916-25G
77360-190EA 90.64 USD
77360-190 77372-154
1-Bromo-1-methoxy-4-phenoxybenzene ≥97%
1-Bromo-1-methoxy-4-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
Storage Temperature: Ambient
CAS Number: 77317-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR