5-Bromo-1-chloro-3-methyl-2-(4-nitrophenoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

147876-5G 147876-25G
77360-184EA 66.95 USD
77360-184 77372-144
5-Bromo-1-chloro-3-methyl-2-(4-nitrophenoxy)benzene ≥97%
5-Bromo-1-chloro-3-methyl-2-(4-nitrophenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrClNO₃
MW: 342.5724 g/mol
Storage Temperature: Ambient
CAS Number: 2921715-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR