2,3-Dibromo-3-(4-bromo-2-methoxyphenyl)-1-(4-bromophenyl)propan-1-one ≥97%

Supplier: AOBChem USA

Warning

147842-5G 147842-25G
77360-182EA 114.33 USD
77360-182 77372-142
2,3-Dibromo-3-(4-bromo-2-methoxyphenyl)-1-(4-bromophenyl)propan-1-one ≥97%
2,3-Dibromo-3-(4-bromo-2-methoxyphenyl)-1-(4-bromophenyl)propan-1-one

Precaución: for Research Use Only

Formula: C₁₆H₁₂Br₄O₂
Storage Temperature: Ambient
CAS Number: 2921880-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR