2-Bromo-4-chloro-5-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

14773-5G 14773-25G
77360-180EA 83.43 USD
77360-180 77372-140
2-Bromo-4-chloro-5-(trifluoromethyl)aniline ≥97%
2-Bromo-4-chloro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrClF₃N
MW: 274.4700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520118
CAS Number: 193090-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR