4-(4-Bromophenoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

147439-5G 147439-25G
77360-174EA 78.28 USD
77360-174 77372-134
4-(4-Bromophenoxy)aniline ≥97%
4-(4-Bromophenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
Storage Temperature: Ambient
CAS Number: 31465-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR