2-Bromo-6-fluoro-4-methylaniline ≥97%

Supplier: AOBChem USA

Warning

14448-5G 14448-25G
77360-148EA 140.08 USD
77360-148 77372-110
2-Bromo-6-fluoro-4-methylaniline ≥97%
2-Bromo-6-fluoro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD07780634
CAS Number: 18349-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR