(E)-2,4,6-trifluorobenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

140011-5G 140011-25G
77360-110EA 60.77 USD
77360-110 77372-076
(E)-2,4,6-trifluorobenzaldehyde oxime ≥97%
(E)-2,4,6-trifluorobenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₄F₃NO
Storage Temperature: Ambient
CAS Number: 2071280-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR