3-Chloro-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Chloro-5-methylphenyl)boronic acid, 3-Chloro-5-methylbenzeneboronic acid

13612-5G 13612-25G
77360-088EA 129.78 USD
77360-088 77372-054
3-Chloro-5-methylphenylboronic acid ≥97%
3-Chloro-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027241
CAS Number: 913836-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR