5-Bromoisoquinolin-1(2H)-one ≥97%

Supplier: AOBChem USA

Warning

129802-5G 129802-25G
77360-040EA 177.16 USD
77360-040 77372-002
5-Bromoisoquinolin-1(2H)-one ≥97%
5-Bromoisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD11226915
CAS Number: 190777-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR