2-Bromo-6-fluoro-5-methylpyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

128650-5G 128650-25G
77360-020EA 123.6 USD
77360-020 77371-986
2-Bromo-6-fluoro-5-methylpyridin-3-ol ≥97%
2-Bromo-6-fluoro-5-methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MW: 206.012 g/mol
Storage Temperature: Ambient
CAS Number: 2921886-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR