1-(3-Chlorophenyl)cycloheptanol ≥97%

Supplier: AOBChem USA

Warning

128250-5G 128250-25G
77360-006EA 63.86 USD
77360-006 77371-982
1-(3-Chlorophenyl)cycloheptanol ≥97%
1-(3-Chlorophenyl)cycloheptanol

Precaución: for Research Use Only

Formula: C₁₃H₁₇ClO
Storage Temperature: Ambient
CAS Number: 39173-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR