2,3-Dibromo-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

126911-5G 126911-25G
77359-990EA 163.77 USD
77359-990 77371-944
2,3-Dibromo-6-methoxybenzoic acid ≥97%
2,3-Dibromo-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₃
MW: 309.939 g/mol
Storage Temperature: Ambient
CAS Number: 2091222-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR