4-Bromo-3,5-difluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

126816-5G 126816-25G
77359-986EA 119.48 USD
77359-986 77371-942
4-Bromo-3,5-difluorobenzonitrile ≥97%
4-Bromo-3,5-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD11845964
CAS Number: 123688-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR