3-Bromo-5-(trifluoromethoxy)benzonitrile ≥97%
Supplier: AOBChem USA
|
Warning
|
126185-5G
126185-25G
77359-978EA
124.63
USD
77359-978
77371-936
3-Bromo-5-(trifluoromethoxy)benzonitrile ≥97%
3-Bromo-5-(trifluoromethoxy)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₃BrF₃NO MW: 266.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08458074 CAS Number: 914635-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...