2-Bromo-3-chloropyridine-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

125016-5G 125016-25G
77359-952EA 164.8 USD
77359-952 77371-924
2-Bromo-3-chloropyridine-4-carboxylic acid ≥97%
2-Bromo-3-chloropyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185789
CAS Number: 1214377-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR