3-Methoxy-1-methylquinolin-1-ium ≥97%

Supplier: AOBChem USA

Warning

124499-5G 124499-25G
77359-938EA 126.69 USD
77359-938 77371-916
3-Methoxy-1-methylquinolin-1-ium ≥97%
3-Methoxy-1-methylquinolin-1-ium

Precaución: for Research Use Only

Formula: C₁₁H₁₂NO
MW: 174.2185 g/mol
Storage Temperature: Ambient
CAS Number: 126189-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR