3,5-Dibromo-2-hydroxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

123249-5G 123249-25G
77359-910EA 75.19 USD
77359-910 77371-900
3,5-Dibromo-2-hydroxybenzonitrile ≥97%
3,5-Dibromo-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Br₂NO
MW: 276.9128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00662354
CAS Number: 40718-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR