2,3-Difluoro(n-hexyloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

12324-5G 12324-25G
77359-908EA 82.4 USD
77359-908 77371-898
2,3-Difluoro(n-hexyloxy)benzene ≥97%
2,3-Difluoro(n-hexyloxy)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD11040406
CAS Number: 121219-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR