1-(Benzofuran-2-yl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

122826-5G 122826-25G
77359-904EA 70.04 USD
77359-904 77371-896
1-(Benzofuran-2-yl)cyclopentanol ≥97%
1-(Benzofuran-2-yl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₂
Storage Temperature: Ambient
CAS Number: 118959-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR